Anti-ART4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51093-T48-50 51093-T48-200 51093-T48-100
103594-682EA 270.87 USD
103594-682 103589-484 103596-116
Anti-ART4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD297 (Catalog#51093-M08H; EDL10575.1; Met1-Lys263). CD297 specific IgG was purified by Mouse CD297 affinity chromatography.

Type: Primary
Antigen: ART4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR