Anti-EGFR Rabbit Monoclonal Antibody [clone:004]

Supplier: Sino Biological
51091-R004-100 51091-R004-50
103596-114EA 150.18 USD
103596-114 103659-912
Anti-EGFR Rabbit Monoclonal Antibody [clone:004]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EGFR / HER1 / ErbB1 (rM EGFR / HER1 / ErbB1; Catalog#51091-M08H; Q01279; Met1-Ser647).

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 004
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR