Anti-IL1R2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51090-RP01-400 51090-RP01-100 51090-RP01-200
103659-906EA 351.49 USD
103659-906 103659-908 103659-910
Anti-IL1R2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL-1R2 / CD121b (Catalog#51090-M08H; P27931; Met1-Glu355). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL1R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR