Anti-ROBO4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51081-T16-200 51081-T16-50 51081-T16-100
103596-102EA 352.03 USD
103596-102 103589-476 103596-100
Anti-ROBO4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse ROBO4 (Catalog#51081-M08H; NP_083059.2; Met1-Glu480). ROBO4 specific IgG was purified by Mouse ROBO4 affinity chromatography.

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR