Anti-FLRT2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51074-T16-50 51074-T16-100 51074-T16-200
103596-094EA 270.87 USD
103596-094 103596-096 103589-474
Anti-FLRT2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse FLRT2 (rM FLRT2; Catalog#51074-M08H; Q8BLU0; Met1-Ser539). FLRT2 specific IgG was purified by Mouse FLRT2 affinity chromatography.

Type: Primary
Antigen: FLRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR