Anti-IL36B Rabbit Polyclonal Antibody

Supplier: Sino Biological
51071-T24-200 51071-T24-100 51071-T24-50
103596-090EA 270.87 USD
103596-090 103596-092 103589-464
Anti-IL36B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL36B/IL1F8 (rM IL36B/IL1F8; Catalog#51071-MNAE; Q9D6Z6; Met1-Lys183). IL36B/IL1F8 specific IgG was purified by Mouse IL36B/IL1F8 affinity chromatography.

Type: Primary
Antigen: IL36B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR