Anti-IL36B Rabbit Polyclonal Antibody

Supplier: Sino Biological

51071-RP02-100 51071-RP02-50 51071-RP02-200
103659-886EA 160.46 USD
103659-886 103596-088 103659-888
Anti-IL36B Rabbit Polyclonal Antibody
Antibodies
Anti-IL36B Antibody

Type: Primary
Antigen: IL36B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR