Anti-IL31 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51070-T16-50 51070-T16-200 51070-T16-100
103589-462EA 106.62 USD
103589-462 103596-084 103596-082
Anti-IL31 Rabbit Polyclonal Antibody
Antibodies
Anti-IL31 Antibody

Type: Primary
Antigen: IL31
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR