Anti-IL31 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51070-RP01-400 51070-RP01-100 51070-RP01-200
103659-880EA 351.49 USD
103659-880 103659-882 103659-878
Anti-IL31 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL31 (Catalog#51070-M08H; NP_083870.1; Met1-Cys163). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR