Anti-IL-5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51068-T16-200 51068-T16-50 51068-T16-100
103596-074EA 214.31 USD
103596-074 103589-458 103596-072
Anti-IL-5 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-5 Antibody

Type: Primary
Antigen: IL-5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR