Anti-IL5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51068-RP01-100 51068-RP01-200 51068-RP01-400
103659-870EA 270.2 USD
103659-870 103659-872 103659-874
Anti-IL5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL-5 / IL5 / Interleukin 5 (rM IL-5 / IL5 / Interleukin 5; Catalog#51068-M08B; P04401; Met1-Gly133). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR