Anti-IL-5 Rabbit Monoclonal Antibody [clone: 121]

Supplier: Sino Biological

51068-R121-50 51068-R121-100
103596-070EA 128.15 USD
103596-070 103659-868
Anti-IL-5 Rabbit Monoclonal Antibody [clone: 121]
Antibodies
Anti-IL-5 Antibody

Type: Primary
Antigen: IL-5
Clonality: Monoclonal
Clone: Clone: 121
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR