Anti-IL17A Rabbit Monoclonal Antibody [clone:215]

Supplier: Sino Biological
51065-R215-50 51065-R215-1 51065-R215-100
103596-064EA 150.18 USD
103596-064 103659-866 103589-454
Anti-IL17A Rabbit Monoclonal Antibody [clone:215]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL17 / IL17a (rM IL17 / IL17a; Catalog#51065-MNAE; Q62386; Thr22-Ala158).

Type: Primary
Antigen: IL17
Clonality: Monoclonal
Clone: 215
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR