Anti-ErbB4 Rabbit Monoclonal Antibody [clone: 049]

Supplier: Sino Biological

51064-R049-100 51064-R049-50
103659-842EA 214.31 USD
103659-842 103596-058
Anti-ErbB4 Rabbit Monoclonal Antibody [clone: 049]
Antibodies
Anti-ErbB4 Antibody

Type: Primary
Antigen: ErbB4
Clonality: Monoclonal
Clone: Clone: 049
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR