Anti-CREG1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51049-T16-100 51049-T16-200 51049-T16-50
103596-048EA 352.03 USD
103596-048 103589-446 103596-046
Anti-CREG1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CREG1 (Catalog#51049-M08H; O88668; Met1-Gln220). CREG1 specific IgG was purified by Mouse CREG1 affinity chromatography.

Type: Primary
Antigen: CREG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR