Anti-CREG1 Rabbit Monoclonal Antibody [clone:111]

Supplier: Sino Biological
51049-R111-100 51049-R111-50
103660-780EA 231.04 USD
103660-780 103596-044
Anti-CREG1 Rabbit Monoclonal Antibody [clone:111]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CREG1 (rM CREG1; Catalog#51049-M08H; O88668; Met1-Gln220).

Type: Primary
Antigen: CREG1
Clonality: Monoclonal
Clone: 111
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR