Anti-CSF2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51048-T16-50 51048-T16-100 51048-T16-200
103596-040EA 352.03 USD
103596-040 103589-444 103596-038
Anti-CSF2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse GM-CSF / CSF2 (rM GM-CSF / CSF2; Catalog#51048-M07H; P01587; Ala18-Lys141). GM-CSF / CSF2 specific IgG was purified by Mouse GM-CSF / CSF2 affinity chromatography.

Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR