Anti-GM-CSF Rabbit Polyclonal Antibody

Supplier: Sino Biological

51048-RP01-200 51048-RP01-400 51048-RP01-100
103659-862EA 160.46 USD
103659-862 103659-864 103659-860
Anti-GM-CSF Rabbit Polyclonal Antibody
Antibodies
Anti-GM-CSF Antibody

Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR