Anti-MOG Rabbit Polyclonal Antibody

Supplier: Sino Biological
51046-T24-200 51046-T24-50 51046-T24-100
103589-442EA 99 USD
103589-442 103596-036 103596-034
Anti-MOG Rabbit Polyclonal Antibody
Anti-MOG Antibody

Type: Primary
Antigen: MOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR