Anti-IGSF8 Rabbit Monoclonal Antibody [clone:002]

Supplier: Sino Biological
51042-R002-100 51042-R002-50
103596-032EA 150.18 USD
103596-032 103659-858
Anti-IGSF8 Rabbit Monoclonal Antibody [clone:002]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IGSF8 (rM IGSF8; Catalog#51042-M08H; AAH48387.1; Met1-Thr577).

Type: Primary
Antigen: IGSF8
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR