Anti-LAIR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51030-T24-100 51030-T24-50 51030-T24-200
103596-026EA 352.03 USD
103596-026 103596-024 103589-438
Anti-LAIR1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LAIR1 (Catalog#51030-M08H; Q8BG84-1; Gln22-Ser133). LAIR1 specific IgG was purified by Mouse LAIR1 affinity chromatography.

Type: Primary
Antigen: LAIR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR