Anti-LAIR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51030-RP01-200 51030-RP01-400 51030-RP01-100
103659-852EA 351.49 USD
103659-852 103659-854 103659-856
Anti-LAIR1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LAIR1 (Catalog#51030-M08H; Q8BG84-1; Gln22-Ser133). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LAIR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR