Anti-LAIR1 Rabbit Monoclonal Antibody [clone:119]

Supplier: Sino Biological
51030-R119-50 51030-R119-100
103659-850EA 271.4 USD
103659-850 103596-022
Anti-LAIR1 Rabbit Monoclonal Antibody [clone:119]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LAIR-1 / CD305 (rM LAIR-1 / CD305; Catalog#51030-M08H; Q8BG84-1; Gln22-Ser133).

Type: Primary
Antigen: LAIR1
Clonality: Monoclonal
Clone: 119
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR