Anti-AKT3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51027-T16-50 51027-T16-100 51027-T16-200
103589-436EA 106.62 USD
103589-436 103596-018 103596-020
Anti-AKT3 Rabbit Polyclonal Antibody
Antibodies
Anti-AKT3 Antibody

Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR