Anti-DPP7 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51019-RP01-400 51019-RP01-200 51019-RP01-100
103660-776EA 562.97 USD
103660-776 103660-774 103660-772
Anti-DPP7 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse DPP7 / DPPII / DPP2 (Catalog#51019-M08H; Q9ET221; Met1-Arg506). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: DPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR