Anti-DPP2 Rabbit Monoclonal Antibody [clone: 035]

Supplier: Sino Biological

51019-R035-100 51019-R035-50
103660-770EA 214.31 USD
103660-770 103596-012
Anti-DPP2 Rabbit Monoclonal Antibody [clone: 035]
Antibodies
Anti-DPP2 Antibody

Type: Primary
Antigen: DPP2
Clonality: Monoclonal
Clone: Clone: 035
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR