Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACPP Rabbit Polyclonal Antibody

Supplier: Sino Biological

51018-T16-100 51018-T16-200 51018-T16-50
103596-008EA 160.46 USD InStock
103596-008 103596-010 103589-432
Anti-ACPP Rabbit Polyclonal Antibody
Antibodies
Anti-Prostatic Acid Phosphatas

Type: Primary
Antigen: ACPP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty