Anti-ACPP Rabbit Monoclonal Antibody [clone: 205]

Supplier: Sino Biological

51018-R205-1 51018-R205-100 51018-R205-50
103589-430EA 1281.54 USD
103589-430 103659-848 103596-006
Anti-ACPP Rabbit Monoclonal Antibody [clone: 205]
Antibodies
Anti-Prostatic Acid Phosphatas

Type: Primary
Antigen: ACPP
Clonality: Monoclonal
Clone: Clone: 205
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR