Anti-ACPP Rabbit Monoclonal Antibody [clone: 202]

Supplier: Sino Biological

51018-R202-50 51018-R202-100
103596-004EA 128.15 USD
103596-004 103659-846
Anti-ACPP Rabbit Monoclonal Antibody [clone: 202]
Antibodies
Anti-Prostatic Acid Phosphatas

Type: Primary
Antigen: ACPP
Clonality: Monoclonal
Clone: Clone: 202
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR