Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACPP Rabbit Monoclonal Antibody [clone: 019]

Supplier: Sino Biological

51018-R019-50 51018-R019-1 51018-R019-100
103596-002EA 128.15 USD InStock
103596-002 103589-428 103659-820
Anti-ACPP Rabbit Monoclonal Antibody [clone: 019]
Antibodies
Anti-Prostatic Acid Phosphatas

Type: Primary
Antigen: ACPP
Clonality: Monoclonal
Clone: Clone: 019
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty