Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL12B Rabbit Polyclonal Antibody

Supplier: Sino Biological

51004-RP01-200 51004-RP01-100 51004-RP01-400
103659-814EA 160.46 USD InStock
103659-814 103659-816 103659-818
Anti-IL12B Rabbit Polyclonal Antibody
Antibodies
Anti-IL12B Antibody

Type: Primary
Antigen: IL12B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty