Anti-ERBB3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51003-RP01-400 51003-RP01-200 51003-RP01-100
103659-808EA 351.49 USD
103659-808 103659-810 103659-806
Anti-ERBB3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse HER3 / ErbB3 (Catalog#51003-M08H; Q61526; Met1-His641). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: HER3 / ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR