Anti-ERBB3 Rabbit Monoclonal Antibody [clone:128]

Supplier: Sino Biological
51003-R128-100 51003-R128-1 51003-R128-50
103589-418EA 1500.59 USD
103589-418 103659-802 103595-984
Anti-ERBB3 Rabbit Monoclonal Antibody [clone:128]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ERBB3 / HER3 (rM ERBB3 / HER3; Catalog#51003-M08H; Q61526; Met1-His641).

Type: Primary
Antigen: HER3 / ERBB3
Clonality: Monoclonal
Clone: 128
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR