Anti-ADAM15 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51001-T16-200 51001-T16-100 51001-T16-50
103595-974EA 214.31 USD
103595-974 103595-972 103589-412
Anti-ADAM15 Rabbit Polyclonal Antibody
Antibodies
Anti-ADAM15 Antibody

Type: Primary
Antigen: ADAM15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR