Anti-ADAM15 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51001-RP01-400 51001-RP01-200 51001-RP01-100
103659-782EA 351.49 USD
103659-782 103659-784 103659-786
Anti-ADAM15 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse ADAM15 (Catalog#51001-M08H; O88839-1; Met1-Thr697). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ADAM15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR