Anti-ADAM15 Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
51001-R001-100 51001-R001-50
103659-780EA 271.4 USD
103659-780 103595-968
Anti-ADAM15 Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ADAM15 (rM ADAM15; Catalog#51001-M08H; O88839-1; Met1-Thr697).

Type: Primary
Antigen: ADAM15
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR