Anti-KDR Rabbit Monoclonal Antibody [clone:004]

Supplier: Sino Biological
50998-R004-100 50998-R004-50
103594-618EA 150.18 USD
103594-618 103659-768
Anti-KDR Rabbit Monoclonal Antibody [clone:004]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VEGFR2 / Flk-1 / CD309 / KDR (rM VEGFR2 / Flk-1 / CD309 / KDR; Catalog#50998-M08H; P35918-1; Met1-Glu762).

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: 004
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR