Anti-KDR Rabbit Monoclonal Antibody [clone:001] APC (Allophycocyanin)

Supplier: Sino Biological
50998-R001-A-25 50998-R001-A-100
103632-352EA 282.8 USD
103632-352 103629-964
Anti-KDR Rabbit Monoclonal Antibody [clone:001] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VEGFR2 / Flk-1 / CD309 / KDR (rM VEGFR2 / Flk-1 / CD309 / KDR; Catalog#50998-M08H; P35918-1; Met1-Glu762) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR