Anti-TH Rabbit Polyclonal Antibody

Supplier: Sino Biological

50997-RP02-100 50997-RP02-200 50997-RP02-50
103659-762EA 160.46 USD
103659-762 103659-764 103594-614
Anti-TH Rabbit Polyclonal Antibody
Antibodies
Anti-Tyrosine Hydroxylase Anti

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR