Anti-TH Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
50997-R001-50 50997-R001-100
103659-754EA 271.1 USD
103659-754 103594-612
Anti-TH Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TH / Tyrosine Hydroxylase (Catalog#50997-M07B; P24529; Pro2-Ser498).

Type: Primary
Antigen: TH
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR