Anti-MFI2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50994-T16-200 50994-T16-100 50994-T16-50
103594-610EA 214.31 USD
103594-610 103594-608 103589-406
Anti-MFI2 Rabbit Polyclonal Antibody
Antibodies
Anti-MFI2 Antibody

Type: Primary
Antigen: MFI2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR