Anti-CPM Rabbit Polyclonal Antibody

Supplier: Sino Biological
50990-RP01-100 50990-RP01-200 50990-RP01-400
103660-766EA 562.97 USD
103660-766 103660-764 103660-762
Anti-CPM Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Carboxypeptidase M / CPM (Catalog#50990-M08H; Q80V42-1; Met1-His422). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CPM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR