Anti-CDH6 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50985-T16-200 50985-T16-100 50985-T16-50
103595-944EA 214.31 USD
103595-944 103595-942 103589-400
Anti-CDH6 Rabbit Polyclonal Antibody
Antibodies
Anti-K Cadherin Antibody

Type: Primary
Antigen: CDH6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR