Anti-C1S1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50979-T16-100 50979-T16-200 50979-T16-50
103595-938EA 270.87 USD
103595-938 103589-398 103595-940
Anti-C1S1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse C1S1 (rM C1S1; Catalog#50979-M08H; AAH18319.1; Met1-Asp688). C1S1 specific IgG was purified by Mouse C1S1 affinity chromatography.

Type: Primary
Antigen: C1S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR