Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ESAM Rabbit Polyclonal Antibody

Supplier: Sino Biological

50977-RP01-100 50977-RP01-400 50977-RP01-200
103660-754EA 106.62 USD InStock
103660-754 103660-756 103659-752
Anti-ESAM Rabbit Polyclonal Antibody
Antibodies
Anti-ESAM Antibody

Type: Primary
Antigen: ESAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty