Anti-ESAM Rabbit Monoclonal Antibody [clone: 110]

Supplier: Sino Biological

50977-R110-100 50977-R110-50
103659-750EA 214.31 USD
103659-750 103595-932
Anti-ESAM Rabbit Monoclonal Antibody [clone: 110]
Antibodies
Anti-ESAM Antibody

Type: Primary
Antigen: ESAM
Clonality: Monoclonal
Clone: Clone: 110
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR