Anti-NBL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50976-T16-50 50976-T16-200 50976-T16-100
103595-928EA 270.87 USD
103595-928 103589-394 103595-930
Anti-NBL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse NBL1 / DAND1 (rM NBL1 / DAND1; Catalog#50976-M08H; Q61477; Met1-Asp178). NBL1 / DAND1 specific IgG was purified by Mouse NBL1 / DAND1 affinity chromatography.

Type: Primary
Antigen: NBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR