Anti-NGFR Rabbit Monoclonal Antibody [clone:117]

Supplier: Sino Biological
50971-R117-100 50971-R117-50
103659-742EA 271.4 USD
103659-742 103595-918
Anti-NGFR Rabbit Monoclonal Antibody [clone:117]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse NGFR / P75 (rM NGFR / P75; Catalog#50971-M08H; Q9Z0W1; Met1-Asn243).

Type: Primary
Antigen: NGFR
Clonality: Monoclonal
Clone: 117
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR