Anti-CPB2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50963-T16-50 50963-T16-100 50963-T16-200
103595-912EA 352.03 USD
103595-912 103595-910 103589-386
Anti-CPB2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Carboxypeptidase B2 / CPB2 (Catalog#50963-M08H; Q9JHH6; Met1-Thr422). Carboxypeptidase B2 / CPB2 specific IgG was purified by Mouse Carboxypeptidase B2 / CPB2 affinity chromatography.

Type: Primary
Antigen: CPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR