Anti-CNDP1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50959-T24-200 50959-T24-100 50959-T24-50
103595-906EA 214.31 USD
103595-906 103595-904 103589-384
Anti-CNDP1 Rabbit Polyclonal Antibody
Antibodies
Anti-CNDP1 Antibody

Type: Primary
Antigen: CNDP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR